Learn More
Cayman Chemical Genipin, Purity: ≥98%, CAS: 6902-77-8, 10 mg
SDP
Catalog No. 502889542
Encompass_Preferred
A protein cross-linking agent isolated from G. jasminoides Ellis; inhibits UCP2 activity; 5 µM genipin increases glucose-stimulated insulin secretion in isolated pancreatic islets chronically exposed to high levels of glucose; induces apoptosis in FaO rat hepatoma cells and human hepatocarcinoma Hep3B cells
Catalog No. 50-288-9542 Supplier Cayman Chemical Supplier No. 1001062210 MG
A protein cross-linking agent isolated from G. jasminoides Ellis; inhibits UCP2 activity; 5 µM genipin increases glucose-stimulated insulin secretion in isolated pancreatic islets chronically exposed to high levels of glucose; induces apoptosis in FaO rat hepatoma cells and human hepatocarcinoma Hep3B cells