Learn More
FUJIFILM BIOSCIENCES INC Genipin, 6902-77-8, MFCD00888600, 5G
Supplier:  FUJIFILM BIOSCIENCES INC 07803021
Encompass_Preferred
Molecular Formula: C11H14O5, Molecular Weight: 226.23, Content: 98.0+% (HPLC), Storage: Room Temperature
Catalog No. NC9449705