Learn More
Medchemexpress LLC (+)-Genipin (Standard) | 6902-77-8 | 226.23 | 50 MG
SDP
Catalog No. 5000386081
Encompass_Preferred
Genipin (Standard) is an analytical standard and natural crosslinking reagent derived from Gardenia jasminoides Ellis fruits. It is intended for research and analytical applications, inhibiting UCP2 (uncoupling protein 2) in cells. This product shares the same bioactivity profile as Genipin and can be utilized for type 2 diabetes research.
  • Protein modulation
  • Antitumor properties
  • Anti-inflammation
  • Immunosuppression
  • Antithrombosis
  • Protection of hippocampal neurons
Catalog No. 50-003-86081 Supplier Medchemexpress LLC Supplier No. HY17389R50MG
Genipin (Standard) is an analytical standard and natural crosslinking reagent derived from Gardenia jasminoides Ellis fruits. It is intended for research and analytical applications, inhibiting UCP2 (uncoupling protein 2) in cells. This product shares the same bioactivity profile as Genipin and can be utilized for type 2 diabetes research.
  • Protein modulation
  • Antitumor properties
  • Anti-inflammation
  • Immunosuppression
  • Antithrombosis
  • Protection of hippocampal neurons