Learn More
Medchemexpress LLC Genipin (Standard) | 6902-77-8 | 97.5% | 226.23 | 5 MG
SDP
Catalog No. 5000383951
Encompass_Preferred
Genipin (Standard) is the analytical standard of Genipin, intended for research and analytical applications. It is a natural crosslinking reagent derived from *Gardenia jasminoides* Ellis fruits and inhibits UCP2 (uncoupling protein 2) in cells. Genipin exhibits various bioactivities, including modulation on proteins, antitumor, anti-inflammation, immunosuppression, antithrombosis, and protection of hippocampal neurons. It is also suitable for type 2 diabetes research.
  • Used as an analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-83951 Supplier Medchemexpress LLC Supplier No. HY17389R5MG
Genipin (Standard) is the analytical standard of Genipin, intended for research and analytical applications. It is a natural crosslinking reagent derived from *Gardenia jasminoides* Ellis fruits and inhibits UCP2 (uncoupling protein 2) in cells. Genipin exhibits various bioactivities, including modulation on proteins, antitumor, anti-inflammation, immunosuppression, antithrombosis, and protection of hippocampal neurons. It is also suitable for type 2 diabetes research.
  • Used as an analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications