Learn More
Apexbio Technology LLC Geniposide 24512-63-8 100mg
SDP
Catalog No. 5000090335
Encompass_Preferred
Geniposide (24512-63-8) is an iridoid glycoside designed to modulate oxidative stress responses by activating and promoting the nuclear translocation of the Nrf2 transcription factor thereby upregulating downstream antioxidant enzyme expression Geniposide exerts its biological activity primarily through Nrf2-mediated pathways In in vitro studies geniposide inhibits tumor cell proliferation and induces apoptosis with reported IC50 values ranging from approximately 20 80 M depending on the cancer cell line Based on these pharmacological properties geniposide holds research potential in neuroprotection cancer biology and metabolic disorder studies including investigation of glucose metabolism and insulin sensitivity
Catalog No. 50-000-90335 Supplier Apexbio Technology LLC Supplier No. N1360100
Geniposide (24512-63-8) is an iridoid glycoside designed to modulate oxidative stress responses by activating and promoting the nuclear translocation of the Nrf2 transcription factor thereby upregulating downstream antioxidant enzyme expression Geniposide exerts its biological activity primarily through Nrf2-mediated pathways In in vitro studies geniposide inhibits tumor cell proliferation and induces apoptosis with reported IC50 values ranging from approximately 20 80 M depending on the cancer cell line Based on these pharmacological properties geniposide holds research potential in neuroprotection cancer biology and metabolic disorder studies including investigation of glucose metabolism and insulin sensitivity