Learn More
Medchemexpress LLC Geniposide | 24512-63-8 | MFCD16036219 | 99.6% | 388.37 g/mol | C17H24O10 | 10 MG
SDP
Catalog No. 5000296972
Encompass_Preferred
Geniposide (standard) is the analytical reference standard of geniposide, provided as a high-purity solid for research and analytical applications. It is intended for use in assay calibration, method development, and chromatographic analysis where an authenticated reference standard is required.
  • High purity suitable for analytical reference work.
  • Provided as a small reference pack for accurate weighing and handling.
  • Appropriate for HPLC, LC-MS, and other chromatographic methods.
  • Supplied as a stable solid with known molecular weight and formula.
  • Includes CAS identifier for unambiguous chemical identification.
Catalog No. 50-002-96972 Supplier Medchemexpress LLC Supplier No. HYN0009R10MG
Geniposide (standard) is the analytical reference standard of geniposide, provided as a high-purity solid for research and analytical applications. It is intended for use in assay calibration, method development, and chromatographic analysis where an authenticated reference standard is required.
  • High purity suitable for analytical reference work.
  • Provided as a small reference pack for accurate weighing and handling.
  • Appropriate for HPLC, LC-MS, and other chromatographic methods.
  • Supplied as a stable solid with known molecular weight and formula.
  • Includes CAS identifier for unambiguous chemical identification.