Learn More
Medchemexpress LLC HY-N0009 50mg Medchemexpress, Geniposide CAS:24512-63-8 Purity:>98%
SDP
Catalog No. 501933350
Encompass_Preferred
Medchemexpress, HY-N0009 50mg Geniposide CAS:24512-63-8 Geniposide is an iridoid glucoside extracted from Gardenia jasminoides Ellis fruits; exhibits a varity of biological activities such as anti-diabetic, antioxidative, antiproliferative and neuroprotective activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3350 Supplier Medchemexpress LLC Supplier No. HYN000950MG
Medchemexpress, HY-N0009 50mg Geniposide CAS:24512-63-8 Geniposide is an iridoid glucoside extracted from Gardenia jasminoides Ellis fruits; exhibits a varity of biological activities such as anti-diabetic, antioxidative, antiproliferative and neuroprotective activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.