Learn More
Medchemexpress LLC Geniposide (Standard) | 24512-63-8 | 98.9% | 388.37 | 50 MG
SDP
Catalog No. 5000428659
Encompass_Preferred
Geniposide (Standard) is the analytical standard of Geniposide. It is intended for research and analytical applications. Geniposide is an iridoid glucoside extracted from *Gardenia jasminoidesEllis* fruits.
  • Exhibits a variety of biological activities such as anti-diabetic, antioxidative, antiproliferative, and neuroprotective activities.
  • Commonly used as an analytical standard in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-28659 Supplier Medchemexpress LLC Supplier No. HYN0009R50MG
Geniposide (Standard) is the analytical standard of Geniposide. It is intended for research and analytical applications. Geniposide is an iridoid glucoside extracted from *Gardenia jasminoidesEllis* fruits.
  • Exhibits a variety of biological activities such as anti-diabetic, antioxidative, antiproliferative, and neuroprotective activities.
  • Commonly used as an analytical standard in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.