Learn More
Medchemexpress LLC Geniposidic acid | 27741-01-1 | 98.9% | 374.34 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN001010MM/1ML
Encompass_Preferred
Geniposidic acid is an orally active FXR modulator and SIRT6 activator. It binds to specific sites on the FXR ligand-binding domain, driving nuclear translocation, coactivator recruitment, and transcription of downstream genes related to bile acid and cholesterol metabolism. It also improves metabolic dysfunction-related fatty liver disease by activating the SIRT6 signaling pathway.
  • Inhibits inflammation.
  • Modulates gut microbiota to alleviate colitis.
  • Can be used in research on drug-induced liver injury.
  • Applicable for inflammatory bowel disease research.
  • Useful for metabolic dysfunction-related fatty liver disease studies.
  • Suitable for research on metabolic dysfunction-related steatohepatitis.
Catalog No. 50-004-28658