Learn More
Cayman Chemical Geniposidic Acid, Purity: ≥98%, CAS: 27741-01-1, 50 mg
SDP
Catalog No. 502940129
Encompass_Preferred
An iridoid glycoside with diverse biological activities; inhibits TPA-induced ear edema in mice by 91.01% when administered topically at 0.1 mg/ear; increases survival rate, attenuates increases in ALT activity and TNF-α levels in serum, as well as hepatic levels of MDA, and prevents decreases in hepatic GSH levels in a mouse model of fulminant hepatic failure induced by D-galactosamine and LPS at 50 mg/kg; decreases systolic blood pressure and heart rate and increases plasma levels of ANP in spontaneously hypertensive rats at 100 mg/kg
Catalog No. 50-294-0129 Supplier Cayman Chemical Supplier No. 2840950 MG
An iridoid glycoside with diverse biological activities; inhibits TPA-induced ear edema in mice by 91.01% when administered topically at 0.1 mg/ear; increases survival rate, attenuates increases in ALT activity and TNF-α levels in serum, as well as hepatic levels of MDA, and prevents decreases in hepatic GSH levels in a mouse model of fulminant hepatic failure induced by D-galactosamine and LPS at 50 mg/kg; decreases systolic blood pressure and heart rate and increases plasma levels of ANP in spontaneously hypertensive rats at 100 mg/kg