Learn More
Apexbio Technology LLC Genistein(Synonyms: 4',5,7-Trihydroxyisoflavone, Genisteol, Prunetol, Sophoricol, Differenol A), 100mg, CAS: 446-72-0.
SDP
Catalog No. 5000060814
Encompass_Preferred
Genistein (CAS 446-72-0) is an isoflavonoid compound that selectively inhibits protein tyrosine kinases enzymes implicated in signaling pathways associated with oncogenic activity and cellular proliferation Genistein inhibits tyrosine kinase activity with an IC50 of approximately 8 M In NIH-3T3 cell assays genistein suppresses epidermal growth factor (EGF)-mediated mitogenesis (IC50 12 M) and insulin-mediated effects (IC50 19 M) additionally inhibiting EGF-induced activation of S6 kinase at a concentration of 6 15 M In animal models oral administration of genistein demonstrates dose-dependent inhibition of prostate adenocarcinoma development highlighting its utility in cancer biology research and potential chemopreventive applications
Catalog No. 50-000-60814 Supplier Apexbio Technology LLC Supplier No. A2198100
Genistein (CAS 446-72-0) is an isoflavonoid compound that selectively inhibits protein tyrosine kinases enzymes implicated in signaling pathways associated with oncogenic activity and cellular proliferation Genistein inhibits tyrosine kinase activity with an IC50 of approximately 8 M In NIH-3T3 cell assays genistein suppresses epidermal growth factor (EGF)-mediated mitogenesis (IC50 12 M) and insulin-mediated effects (IC50 19 M) additionally inhibiting EGF-induced activation of S6 kinase at a concentration of 6 15 M In animal models oral administration of genistein demonstrates dose-dependent inhibition of prostate adenocarcinoma development highlighting its utility in cancer biology research and potential chemopreventive applications