Learn More
AdipoGen Genistein (100 mg)
SDP
Supplier:  AdipoGen CDXG0210M100
Encompass_Preferred
Chemical. CAS: 446-72-0. Formula: C15H10O5. MW: 270.24. Genistein is an isoflavonoid phytoestrogen that has been found in soybeans and has kinase inhibitory, anticancer, pro-cancer and hepatoprotective properties. It is a cell-permeable, reversible, substrate competitive tyrosine kinase inhibitor (including EGFR phosphorylation), implicated in almost all cell growth and proliferation signal cascades. Genistein inhibits proliferation and induces cell cycle arrest and apoptosis in a variety of cancer cells. It is antiangiogenic agent, down-regulates the transcription of genes involved in controlling angiogenesis. On a molecular level it also inhibits mammalian DNA topoisomerase II and DNA methyltransferase. Genistein has also antidiabetic, anti-inflammatory, antimicrobial and antioxidant properties. It is an alpha-glucosidase inhibitor, PPAR activator, GLUT4-mediated glucose uptake inhibitor, MALT1 inhibitor and acts as an agonist at the GPR30 receptor.
Catalog No. 50-205-4471