Learn More
Medchemexpress LLC Genistein | 446-72-0 | 99.8% | 1 ML
SDP
Catalog No. 5000354466
Encompass_Preferred
Genistein is a soy isoflavone and a multiple tyrosine kinase inhibitor (e.g., EGFR). It acts as a chemotherapeutic agent against different types of cancer by altering apoptosis, the cell cycle, and angiogenesis, and by inhibiting metastasis. This product is for research use only.
  • Soy isoflavone
  • Multiple tyrosine kinase inhibitor (e.g., EGFR)
  • Chemotherapeutic agent against various cancers
  • Modulates apoptosis and cell cycle
  • Inhibits angiogenesis and metastasis
  • Targets EGFR
  • IC50 of 0.6 μM (cell assay)
  • Provided as 10 mM solution in 1 mL DMSO
Catalog No. 50-003-54466 Supplier Medchemexpress LLC Supplier No. HY1459610MM/1ML
Genistein is a soy isoflavone and a multiple tyrosine kinase inhibitor (e.g., EGFR). It acts as a chemotherapeutic agent against different types of cancer by altering apoptosis, the cell cycle, and angiogenesis, and by inhibiting metastasis. This product is for research use only.
  • Soy isoflavone
  • Multiple tyrosine kinase inhibitor (e.g., EGFR)
  • Chemotherapeutic agent against various cancers
  • Modulates apoptosis and cell cycle
  • Inhibits angiogenesis and metastasis
  • Targets EGFR
  • IC50 of 0.6 μM (cell assay)
  • Provided as 10 mM solution in 1 mL DMSO