Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Genistein | 446-72-0 | MFCD00016952 | C15H10O5 | 10MG
SDP

Catalog No. 5000259596
Encompass_Preferred

Genistein analytical standard is a soy isoflavone reference material supplied for research and analytical use. The material has confirmed identity (CAS 446-72-0), formula C15H10O5, and molecular weight ~270.24 g/mol. It is supplied as a powder with documented stability guidelines and accompanying documentation to support analytical workflows.

  • Analytical reference suitable for method validation and quality control.
  • High purity: 99.3% as stated on supplier documentation.
  • Documented chemical identity and properties (CAS 446-72-0, C15H10O5).
  • Recommended cold storage for long-term stability.
  • Provided with certificate of analysis and handling documentation.

Catalog No. 50-002-59596 Supplier Medchemexpress LLC Supplier No. HY14596R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Genistein analytical standard is a soy isoflavone reference material supplied for research and analytical use. The material has confirmed identity (CAS 446-72-0), formula C15H10O5, and molecular weight ~270.24 g/mol. It is supplied as a powder with documented stability guidelines and accompanying documentation to support analytical workflows.

  • Analytical reference suitable for method validation and quality control.
  • High purity: 99.3% as stated on supplier documentation.
  • Documented chemical identity and properties (CAS 446-72-0, C15H10O5).
  • Recommended cold storage for long-term stability.
  • Provided with certificate of analysis and handling documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.