Learn More
Selleck Chemical LLC Genistein S1342-10mM/1mL
SDP
Catalog No. 503110358
Encompass_Preferred
Genistein a phytoestrogen found in soy products is a highly specific inhibitor of protein tyrosine kinase (PTK) which blocks the mitogenic effect mediated by EGF on NIH-3T3 cells with IC50 of 12 M
Catalog No. 50-311-0358 Supplier Selleck Chemical LLC Supplier No. S134210MM/1ML
Genistein a phytoestrogen found in soy products is a highly specific inhibitor of protein tyrosine kinase (PTK) which blocks the mitogenic effect mediated by EGF on NIH-3T3 cells with IC50 of 12 M