Learn More
Medchemexpress LLC Genistein 7-β-D-Glucuronide | 38482-81-4 | 98.8% | 446.36 | 1 MG
SDP
Catalog No. 5000350004
Encompass_Preferred
Genistein 7-β-D-Glucuronide is a natural product that functions as an inhibitor of tubulogenesis and endothelial migration. It is intended for research use only.
  • Inhibits tubulogenesis
  • Inhibits endothelial migration
  • Natural product
  • Targets vascular endothelial growth factor receptor (VEGFR)
  • Relevant to metabolic or endocrine disease research
Catalog No. 50-003-50004 Supplier Medchemexpress LLC Supplier No. HY1379671MG
Genistein 7-β-D-Glucuronide is a natural product that functions as an inhibitor of tubulogenesis and endothelial migration. It is intended for research use only.
  • Inhibits tubulogenesis
  • Inhibits endothelial migration
  • Natural product
  • Targets vascular endothelial growth factor receptor (VEGFR)
  • Relevant to metabolic or endocrine disease research