Learn More
Medchemexpress LLC Genistein 7-sulfate sodium | 99.9% | 372.28 | 1 MG
SDP
Catalog No. 5000349021
Encompass_Preferred
Genistein 7-sulfate sodium is a metabolite of genistein. It has the ability to reduce the activity of estrogen agonists in MCF-7 cells and can also promote the growth of MCF-7 cells at concentrations of 10 μM and above.
  • Metabolite of genistein
  • Reduces activity of estrogen agonists in MCF-7 cells
  • Promotes growth of MCF-7 cells at concentrations of 10 μM and above
  • Used in research on effects on estrogen agonists and MCF-7 cell growth
Catalog No. 50-003-49021 Supplier Medchemexpress LLC Supplier No. HY137099A1MG
Genistein 7-sulfate sodium is a metabolite of genistein. It has the ability to reduce the activity of estrogen agonists in MCF-7 cells and can also promote the growth of MCF-7 cells at concentrations of 10 μM and above.
  • Metabolite of genistein
  • Reduces activity of estrogen agonists in MCF-7 cells
  • Promotes growth of MCF-7 cells at concentrations of 10 μM and above
  • Used in research on effects on estrogen agonists and MCF-7 cell growth