Learn More
Medchemexpress LLC HY-N6882 1mg Medchemexpress, Genistein 8-c-glucoside CAS:66026-80-0 Purity:>98%
SDP
Catalog No. 501970479
Encompass_Preferred
Medchemexpress, HY-N6882 1mg Genistein 8-c-glucoside CAS:66026-80-0 Genistein 8-c-glucoside (G8CG) is a natural glucoside isolated from flowers of Lupinus luteus L. Genistein 8-c-glucoside induces mitochondrial membrane depolarization and induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0479 Supplier Medchemexpress LLC Supplier No. HYN68821MG
Medchemexpress, HY-N6882 1mg Genistein 8-c-glucoside CAS:66026-80-0 Genistein 8-c-glucoside (G8CG) is a natural glucoside isolated from flowers of Lupinus luteus L. Genistein 8-c-glucoside induces mitochondrial membrane depolarization and induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.