Learn More
Cayman Chemical Genistein-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 187960-08-3, 1 mg
SDP
Catalog No. 502966038
Encompass_Preferred
An internal standard for the quantification of genistein by GC- or LC-MS
Catalog No. 50-296-6038 Supplier Cayman Chemical Supplier No. 182441 MG
An internal standard for the quantification of genistein by GC- or LC-MS