Learn More
Cayman Chemical GenIsteIn-d4 5mg
SDP
Catalog No. 502968144
Encompass_Preferred
An internal standard for the quantification of genistein by GC- or LC-MS
Catalog No. 50-296-8144 Supplier Cayman Chemical Supplier No. 182445 MG
Add to Cart
Add to Cart
An internal standard for the quantification of genistein by GC- or LC-MS