Learn More
Selleck Chemical LLC Genistein 100mg 446-72-0
SDP
Catalog No. 507860
Encompass_Preferred
Genistein, a phytoestrogen found in soy products, is a highly specific inhibitor of protein tyrosine kinase (PTK) which blocks the mitogenic effect mediated by EGF on NIH-3T3 cells with IC50 of 12?M or by insulin with IC50 of 19 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-786-0 Supplier Selleck Chemical LLC Supplier No. S1342100MG
Genistein, a phytoestrogen found in soy products, is a highly specific inhibitor of protein tyrosine kinase (PTK) which blocks the mitogenic effect mediated by EGF on NIH-3T3 cells with IC50 of 12?M or by insulin with IC50 of 19 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.