Learn More
Abcam Genistein, Protein tyrosine kinase inhibitor, 100MG
Catalog No. 502707870
Encompass_Preferred
MW 270.24 Da, Purity >99%. Phytoestrogen found in soy beans. Anticancer agent. Inihibits tyrosine protein kinase and DNA topoisomerase as well as possessing antioxidant and cell cycle inhibitor activity. Endocytosis inhibitor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7870 Supplier Abcam Supplier No. AB120112100MG
MW 270.24 Da, Purity >99%. Phytoestrogen found in soy beans. Anticancer agent. Inihibits tyrosine protein kinase and DNA topoisomerase as well as possessing antioxidant and cell cycle inhibitor activity. Endocytosis inhibitor.
The product is subject to the following: Abcam Restricted Use Statement