Learn More
Abcam Genistein, Protein tyrosine kinase inhibitor, 50MG
Catalog No. 502707868
Encompass_Preferred
MW 270.24 Da, Purity >99%. Phytoestrogen found in soy beans. Anticancer agent. Inihibits tyrosine protein kinase and DNA topoisomerase as well as possessing antioxidant and cell cycle inhibitor activity. Endocytosis inhibitor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7868 Supplier Abcam Supplier No. AB12011250MG
MW 270.24 Da, Purity >99%. Phytoestrogen found in soy beans. Anticancer agent. Inihibits tyrosine protein kinase and DNA topoisomerase as well as possessing antioxidant and cell cycle inhibitor activity. Endocytosis inhibitor.
The product is subject to the following: Abcam Restricted Use Statement