Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Genistein (Standard) | 446-72-0 | MFCD00016952 | 98.7% | C₁₅H₁₀O₅ | 25 MG
SDP

Catalog No. 5000353503
Encompass_Preferred

Genistein (Standard) is the analytical standard of Genistein, a soy isoflavone. Intended for research and analytical applications, it acts as a chemotherapeutic agent by inhibiting multiple tyrosine kinases (e.g., EGFR), altering apoptosis, cell cycle, angiogenesis, and inhibiting metastasis. Commonly used in HPLC, GC, and MS experiments, it is a light yellow to yellow solid, with a molecular weight of 270.24. Classified as a flavonoid and phenol, its sources include plants and human gut microbiota metabolites.

  • Analytical standard for research and analytical applications
  • Inhibits multiple tyrosine kinases; acts as chemotherapeutic agent
  • Modulates apoptosis, cell cycle, angiogenesis; inhibits metastasis
  • Used in HPLC, GC, and MS experiments
  • Molecular weight: 270.24
  • Appearance: Light yellow to yellow solid
  • Classified as flavonoid and phenol
  • Derived from plants and human gut microbiota metabolites

Catalog No. 50-003-53503 Supplier Medchemexpress LLC Supplier No. 25MGHY14596R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Genistein (Standard) is the analytical standard of Genistein, a soy isoflavone. Intended for research and analytical applications, it acts as a chemotherapeutic agent by inhibiting multiple tyrosine kinases (e.g., EGFR), altering apoptosis, cell cycle, angiogenesis, and inhibiting metastasis. Commonly used in HPLC, GC, and MS experiments, it is a light yellow to yellow solid, with a molecular weight of 270.24. Classified as a flavonoid and phenol, its sources include plants and human gut microbiota metabolites.

  • Analytical standard for research and analytical applications
  • Inhibits multiple tyrosine kinases; acts as chemotherapeutic agent
  • Modulates apoptosis, cell cycle, angiogenesis; inhibits metastasis
  • Used in HPLC, GC, and MS experiments
  • Molecular weight: 270.24
  • Appearance: Light yellow to yellow solid
  • Classified as flavonoid and phenol
  • Derived from plants and human gut microbiota metabolites

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.