Learn More
Medchemexpress LLC Genistein (Standard) | 446-72-0 | MFCD00016952 | 98.7% | C₁₅H₁₀O₅ | 50 MG
SDP
Catalog No. 5000353316
Encompass_Preferred
Genistein (Standard) is an analytical standard and soy isoflavone. It acts as a multiple tyrosine kinases inhibitor, used in chemotherapeutic research to alter apoptosis, cell cycle, angiogenesis, and inhibit metastasis. This product is intended for research and analytical applications.
  • Reference standard for assay
  • Used in qualitative research
  • Used in quantitative research
  • Used in methodological research experiments
  • Compatible with HPLC, GC, and MS
Catalog No. 50-003-53316 Supplier Medchemexpress LLC Supplier No. HY14596R50MG
Genistein (Standard) is an analytical standard and soy isoflavone. It acts as a multiple tyrosine kinases inhibitor, used in chemotherapeutic research to alter apoptosis, cell cycle, angiogenesis, and inhibit metastasis. This product is intended for research and analytical applications.
  • Reference standard for assay
  • Used in qualitative research
  • Used in quantitative research
  • Used in methodological research experiments
  • Compatible with HPLC, GC, and MS