Learn More
Enzo Life Sciences Genistein (synthetic) (25mg). CAS: 446-72-0
SDP
Supplier:  Enzo Life Sciences ALX350006M025
Encompass_Preferred
Tyrosine protein kinase inhibitor. Inhibits phosphorylation of EGFR kinase. Inhibits tumor cell proliferation and induces tumor cell differentiation. Inhibits topoisomerase II activity in vivo. Produces cell cycle arrest and apoptosis. Potent inhibitor of α-glucosidase, IC50 = 50nM.Direct inhibitor of insulin-induced glucose uptake in adipocytes (IC50=20µM). Alternative name: 4',5,7-Trihydroxyisoflavone. Purity: ≥98% (HPLC). Appearance: Creamy off-white solid. Solubility: Soluble in DMSO (30mg/ml), 100% ethanol (4mg/ml) or dilute aqueous base; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8682