Learn More
Sigma Aldrich Fine Chemicals Biosciences Genistein synthetic 98 HPL5MG
Catalog No. 502897604
Encompass
Genistein is an isoflavonoid present among Leguminosae family of plants. It is structurally similar to estrogen (estradiol-17).
Catalog No. 50-289-7604 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G66495MG
Genistein is an isoflavonoid present among Leguminosae family of plants. It is structurally similar to estrogen (estradiol-17).