Learn More
Sigma Aldrich Fine Chemicals Biosciences Genistein synthtic 98 HPL100MG
Catalog No. 502902162
Encompass
Genistein is an isoflavonoid present among Leguminosae family of plants. It is structurally similar to estrogen (estradiol-17).
Catalog No. 50-290-2162 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G6649100MG
Genistein is an isoflavonoid present among Leguminosae family of plants. It is structurally similar to estrogen (estradiol-17).