Learn More
Enzo Life Sciences Genistin (10mg). CAS: 529-59-9
SDP
Supplier:  Enzo Life Sciences ALX350247M010
Encompass_Preferred
Glucoside of genistein (ALX-350-006) found in soy beans. Useful as a negative control for genistein and other tyrosine kinase inhibitors. Selective inhibitor of terminal deoxyribonucleotidyltransferase (TdT). Displays antioxidant and anticarcinogenic properties. Alternative name: Genistein-7-O-glucoside. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in DMSO or dilute aqueous base; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8801