Learn More
Medchemexpress LLC Genistin | 529-59-9 | 432.38 | 1 ML
SDP
Catalog No. 5000388370
Encompass_Preferred
Genistin, an isoflavone from the phytoestrogen family, is a potent anti-adipogenic and anti-lipogenic agent. It attenuates cellular growth and promotes apoptotic cell death in breast cancer cells by modulating the ERalpha signaling pathway. It also effectively down-modulates ER nuclear translocation and DNA binding activity, inducing apoptosis and suppressing oncogenic markers in MCF-7 cells.
  • Potent anti-adipogenic and anti-lipogenic agent
  • Attenuates cellular growth and promotes apoptosis
  • Modulates ERalpha signaling pathway
  • Down-modulates ER nuclear translocation and DNA binding activity
  • Induces apoptosis and suppresses oncogenic markers
Catalog No. 50-003-88370 Supplier Medchemexpress LLC Supplier No. HYN059510MM/1ML
Genistin, an isoflavone from the phytoestrogen family, is a potent anti-adipogenic and anti-lipogenic agent. It attenuates cellular growth and promotes apoptotic cell death in breast cancer cells by modulating the ERalpha signaling pathway. It also effectively down-modulates ER nuclear translocation and DNA binding activity, inducing apoptosis and suppressing oncogenic markers in MCF-7 cells.
  • Potent anti-adipogenic and anti-lipogenic agent
  • Attenuates cellular growth and promotes apoptosis
  • Modulates ERalpha signaling pathway
  • Down-modulates ER nuclear translocation and DNA binding activity
  • Induces apoptosis and suppresses oncogenic markers