Learn More
Medchemexpress LLC Genistin | 529-59-9 | 432.38 | 100 MG
SDP
Catalog No. 5000389031
Encompass_Preferred
Genistin (Genistine), an isoflavone belonging to the phytoestrogen family, is a potent anti-adipogenic and anti-lipogenic agent. Genistin attenuates cellular growth and promotes apoptotic cell death breast cancer cells through modulation of the ERalpha signaling pathway.
  • An isoflavone belonging to the phytoestrogen family.
  • Potent anti-adipogenic and anti-lipogenic agent.
  • Attenuates cellular growth.
  • Promotes apoptotic cell death in breast cancer cells through modulation of the ERalpha signaling pathway.
Catalog No. 50-003-89031 Supplier Medchemexpress LLC Supplier No. HYN0595100MG
Genistin (Genistine), an isoflavone belonging to the phytoestrogen family, is a potent anti-adipogenic and anti-lipogenic agent. Genistin attenuates cellular growth and promotes apoptotic cell death breast cancer cells through modulation of the ERalpha signaling pathway.
  • An isoflavone belonging to the phytoestrogen family.
  • Potent anti-adipogenic and anti-lipogenic agent.
  • Attenuates cellular growth.
  • Promotes apoptotic cell death in breast cancer cells through modulation of the ERalpha signaling pathway.