Learn More
Medchemexpress LLC Genistin | 529-59-9 | 432.38 | 200 MG
SDP
Catalog No. 5000388628
Encompass_Preferred
Genistin (Genistine), an isoflavone belonging to the phytoestrogen family, is a potent anti-adipogenic and anti-lipogenic agent. It attenuates cellular growth and promotes apoptotic cell death in breast cancer cells through modulation of the ERalpha signaling pathway.
  • Negatively regulates ERα.
  • Down-modulates ER nuclear translocation and DNA binding activity in breast cancer cells.
  • Induces apoptosis and suppresses levels of oncogenic markers in MCF-7 cells.
Catalog No. 50-003-88628 Supplier Medchemexpress LLC Supplier No. HYN0595200MG
Genistin (Genistine), an isoflavone belonging to the phytoestrogen family, is a potent anti-adipogenic and anti-lipogenic agent. It attenuates cellular growth and promotes apoptotic cell death in breast cancer cells through modulation of the ERalpha signaling pathway.
  • Negatively regulates ERα.
  • Down-modulates ER nuclear translocation and DNA binding activity in breast cancer cells.
  • Induces apoptosis and suppresses levels of oncogenic markers in MCF-7 cells.