Learn More
Medchemexpress LLC Genistin | 529-59-9 | 432.38 | 50 MG
SDP
Catalog No. 5000388629
Encompass_Preferred
Genistin is an isoflavone from the phytoestrogen family. It functions as a potent anti-adipogenic and anti-lipogenic agent. This compound has demonstrated the ability to attenuate cellular growth and promote apoptotic cell death in breast cancer cells by modulating the ERalpha signaling pathway.
  • Potent anti-adipogenic and anti-lipogenic agent
  • Attenuates cellular growth
  • Promotes apoptotic cell death in breast cancer cells
  • Modulates the ERalpha signaling pathway
Catalog No. 50-003-88629 Supplier Medchemexpress LLC Supplier No. HYN059550MG
Genistin is an isoflavone from the phytoestrogen family. It functions as a potent anti-adipogenic and anti-lipogenic agent. This compound has demonstrated the ability to attenuate cellular growth and promote apoptotic cell death in breast cancer cells by modulating the ERalpha signaling pathway.
  • Potent anti-adipogenic and anti-lipogenic agent
  • Attenuates cellular growth
  • Promotes apoptotic cell death in breast cancer cells
  • Modulates the ERalpha signaling pathway