Learn More
Peak Scientific Instruments Ltd Genius XE 35 Nitrogen Generator Produces up to 35L/min of high purity N2 for mass spectrometry
Supplier: Peak Scientific Instruments Ltd 3300252
Variable flow up to 35 LPM
Variable pressure up to 116 psi
Multi-Stage Purification™ producing analytical grade nitrogen gas up to 99.5% purity
2 year comprehensive manufacturer’s warranty*
Touch-screen full colour user interface for ease of operation
ECO (Electronic Compressor Optimisation™) technology for low energy consumption and compressor durability
Next-generation high performance premium compressors, engineered exclusively for Genius XE
Smaller & quieter than previous generation of Genius
On-board service scheduling and system diagnostics