Learn More
Peak Scientific Instruments Ltd Genius XE 70 Nitrogen Generator Produces up to 70L/min of high purity N2 for mass spectrometry
SDP
Catalog No. NC1583509
Encompass_Preferred
Variable flow up to 70 L/min & pressure up to 116 psiMulti-Stage Purification™ producing analytical grade nitrogen gas up to 99.5% purity2 year comprehensive manufacturer’s warranty*Touch-screen full colour user interface for ease of operationECO (Electronic Compressor Optimisation™) technology for low energy consumption and compressor durabilityNext-generation high performance premium compressors, engineered exclusively for Genius XESmaller & quieter than previous generation of GeniusOn-board service scheduling and system diagnosticsAdditional compressor capacity for lower flow applications
Catalog No. NC1583509 Supplier Peak Scientific Instruments Ltd Supplier No. 3300253
Add to Cart
Add to Cart
Request a Quote
Variable flow up to 70 L/min & pressure up to 116 psiMulti-Stage Purification™ producing analytical grade nitrogen gas up to 99.5% purity2 year comprehensive manufacturer’s warranty*Touch-screen full colour user interface for ease of operationECO (Electronic Compressor Optimisation™) technology for low energy consumption and compressor durabilityNext-generation high performance premium compressors, engineered exclusively for Genius XESmaller & quieter than previous generation of GeniusOn-board service scheduling and system diagnosticsAdditional compressor capacity for lower flow applications