Learn More
SignaGen Laboratories GENJET PLUS DNA REAGENT 1ML
Catalog No. 504779
Encompass
GenJet Plus DNA In Vitro Tranfection Reagent is enhanced version of GenJet DNA Transfection Reagent (Ver. II) with addition of a booster peptide, giving rise to up to 10 times better efficiency than its previous version on variety of mammalian cells.
Catalog No. 50-477-9 Supplier SignaGen Laboratories Supplier No. SL100499
GenJet Plus DNA In Vitro Tranfection Reagent is enhanced version of GenJet DNA Transfection Reagent (Ver. II) with addition of a booster peptide, giving rise to up to 10 times better efficiency than its previous version on variety of mammalian cells.