Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amsbio LLC (formerly Genlantis) GenePORTER™ 3000 Reagent

Catalog No. T203007
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T203007 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T203007 Supplier Amsbio LLC (formerly Genlantis) Supplier No. AMS.T203007
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Provides high transfection efficiency for hard to transfect cells

The GenePORTER 3000™ Transfection Reagent is a proprietary cationic lipid formulation that utilizes Advanced Carrier Enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity.

  • Designed specifically for difficult-to-transfect cells
  • Rapid protocol: Optimized for same day transfection
  • Superior performance in both serum-free and serum-containing media

Specifications

For Use With (Application) Proprietary cationic Lipid that utilizes advanced conjugation enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity
Includes 0.75mL GenePORTER™ 3000 reagent, 6mL GP3K diluents
Quantity 0.75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.