Learn More
Amsbio LLC (formerly Genlantis) GenePORTER™ Transfection Reagents and Kits
Catalog No. T201015
Click to view available options
Quantity:
1.5 mL
Catalog No. Quantity
T201015 1.5 mL
1 options
Catalog No. T201015 Supplier Amsbio LLC (formerly Genlantis) Supplier No. AMS.T201015
High-efficiency, serum-free transfection of a broad spectrum of adherent and suspension cells
  • Low cytotoxicity
  • Ideal for delivery of oligonucleotides
  • Unique lipid formulation that allows efficient serum-free transfection of a broad spectrum of adherent and suspension cells
Specifications
For Use With (Application) Transfection of broad spectrum of adherent and suspension cells, effective DNA complexing, cytoplasmic translocation and subsequent nuclear release of plasmid DNA and DNA Oligonucleotides
Includes Cationic Lipid, Hydration Buffer with Neutral Lipid Dioleoyl Phosphatidylethanolamine
Quantity 1.5 mL