Learn More
SignaGen Laboratories GENMUTE REAGENT 1ML
Catalog No. 504858
Encompass
GenMute siRNA Transfection Reagent, formulated by PDCC technology, is a novel biodegradable non-liposomal siRNA & DNA delivery tool with exceptional silencing at 1.0 nM siRNA.
Catalog No. 50-485-8 Supplier SignaGen Laboratories Supplier No. SL100568
GenMute siRNA Transfection Reagent, formulated by PDCC technology, is a novel biodegradable non-liposomal siRNA & DNA delivery tool with exceptional silencing at 1.0 nM siRNA.