Learn More
Sigma Aldrich Fine Chemicals Biosciences GenomePlexR Complete Whole
Catalog No. 502914681
Encompass
GenomePlex Complete Whole Genome Amplification Kit utilizes a proprietary technology based on random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable library molecules flanked by universal priming sites. WGA is achieved by PCR amplification of the library molecules using universal oligonucleotide primers.
Catalog No. 50-291-4681 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WGA2500RXN
Add to Cart
Add to Cart
Request a Quote
GenomePlex Complete Whole Genome Amplification Kit utilizes a proprietary technology based on random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable library molecules flanked by universal priming sites. WGA is achieved by PCR amplification of the library molecules using universal oligonucleotide primers.