Learn More
Sigma Aldrich Fine Chemicals Biosciences GenomePlexR Single Cell Wh
Catalog No. 502914693
Encompass
GenomePlex Single Cell Whole Genome Amplification Kit utilizes a proprietary technology based on random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable library molecules flanked by universal priming sites. WGA is achieved by PCR amplification of the library molecules using universal oligonucleotide primers.
Catalog No. 50-291-4693 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WGA450RXN
GenomePlex Single Cell Whole Genome Amplification Kit utilizes a proprietary technology based on random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable library molecules flanked by universal priming sites. WGA is achieved by PCR amplification of the library molecules using universal oligonucleotide primers.