Learn More
Sigma Aldrich Fine Chemicals Biosciences GenomePlexR WGA Reamplific
Catalog No. 502904127
Encompass
GenomePlex WGA Reamplification Kit utilizes a proprietary amplification method that is based on random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable library molecules flanked by universal priming sites. WGA is achieved by PCR amplification of the library molecules using universal oligonucleotide primers.
Catalog No. 50-290-4127 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WGA350RXN
Add to Cart
Add to Cart
GenomePlex WGA Reamplification Kit utilizes a proprietary amplification method that is based on random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable library molecules flanked by universal priming sites. WGA is achieved by PCR amplification of the library molecules using universal oligonucleotide primers.