Learn More
Zymo Research Corporation Genomic DNA Clean & Concentrator™ Kit-5 (4 x 96 Preps)
SDP
Catalog No. 501251424
Encompass_Preferred
The ZR-96 Genomic DNA Clean & Concentrator-5 (DCC) is for high throughput recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, (wga) DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied ZymoSpin™ I-96-XL Plate. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions,crude plasmid preparations, cDNA synthesis, etc.
Catalog No. 50-125-1424 Supplier Zymo Research Corporation Supplier No. D4067
The ZR-96 Genomic DNA Clean & Concentrator-5 (DCC) is for high throughput recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, (wga) DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied ZymoSpin™ I-96-XL Plate. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions,crude plasmid preparations, cDNA synthesis, etc.