Learn More
Zymo Research Corporation Genomic DNA Clean & Concentrator™ Kit-25 (100 Preps)
SDP
Catalog No. NC0760907
Encompass_Preferred
There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied ZymoSpin™ Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions,crude plasmid preparations, cDNA synthesis, etc.
Catalog No. NC0760907 Supplier Zymo Research Corporation Supplier No. D4065
There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied ZymoSpin™ Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions,crude plasmid preparations, cDNA synthesis, etc.