Learn More
Research Products International Corp Genomic DNA Clean & Concentrator-10 Kit, 25 Preps
SDP
Catalog No. 501977877
Encompass
The Genomic DNA Clean & Concentrator-10 (gDCC) is a gDNA clean up kit that provides quick, 5 minute recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, WGA DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied Zymo-Spin Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. This gDNA clean up kit is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, etc.
Quick, 5-minute clean-up of large-sized DNA from any enzymatic reaction or impure preparation without messy precipitations.
Catalog No. 50-197-7877 Supplier Research Products International Corp Supplier No. ZD4010
The Genomic DNA Clean & Concentrator-10 (gDCC) is a gDNA clean up kit that provides quick, 5 minute recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, WGA DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied Zymo-Spin Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. This gDNA clean up kit is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, etc.
Quick, 5-minute clean-up of large-sized DNA from any enzymatic reaction or impure preparation without messy precipitations.