Learn More
Research Products International Corp Genomic DNA Clean & Concentrator Kit-25, 100 Preps
SDP
Supplier:  Research Products International Corp ZD4065
Encompass
The Genomic DNA Clean & Concentrator-25 (DCC) is for quick (5 minute) recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, (wga)DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Bufferto a sample and then transfer the mixture to the supplied Zymo-Spin Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, etc.
Quick (5 minute) spin column recovery of large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, (wga)DNA, etc.) from anyenzymatic reaction or impure preparation (e.g., Proteinase K digestion). No messy precipitations!
Catalog No. 50-197-7275