Learn More
Research Products International Corp Genomic DNA Clean & Concentrator Kit (100 Preps)
SDP
Catalog No. 501977272
Encompass
The Genomic DNA Clean & Concentrator-10 (gDCC) is a gDNA clean up kit that provides quick, 5 minute recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, WGA DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied Zymo-Spin Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. This gDNA clean up kit is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, etc.
Quick, 5-minute clean-up of large-sized DNA from any enzymatic reaction or impure preparation without messy precipitations.
Catalog No. 50-197-7272 Supplier Research Products International Corp Supplier No. ZD4011
The Genomic DNA Clean & Concentrator-10 (gDCC) is a gDNA clean up kit that provides quick, 5 minute recovery of ultra-pure, large-sized DNA (e.g., genomic, mitochondrial, plasmid (BAC/PAC), viral, phage, WGA DNA, etc.) from any enzymatic reaction or impure preparation (e.g., Proteinase K digestion). There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied Zymo-Spin Column. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. This gDNA clean up kit is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, etc.
Quick, 5-minute clean-up of large-sized DNA from any enzymatic reaction or impure preparation without messy precipitations.