Learn More
Biochain Institute Inc Control Genomic DNA - Human Male, 100 ug/PK
SDP
Catalog No. 50181348
Encompass_Preferred
BioChain's control genomic DNAs are isolated from tissues using a proprietary modified guanidine thiocyanate technique. The quality and purity of genomic DNA are tested by spectrophotometer and electrophoresis. A260/280 is between 1.8 and 2.0, and A260/230 is >;2.0. Contamination of RNA, polysaccharides and proteoglycans has been effectively eliminated.
Catalog No. 50-181-348 Supplier Biochain Institute Inc Supplier No. D1234999G01
BioChain's control genomic DNAs are isolated from tissues using a proprietary modified guanidine thiocyanate technique. The quality and purity of genomic DNA are tested by spectrophotometer and electrophoresis. A260/280 is between 1.8 and 2.0, and A260/230 is >;2.0. Contamination of RNA, polysaccharides and proteoglycans has been effectively eliminated.