Learn More
TRIBIOSCIENCE INC Genomic DNA Extraction from Cells and Tissues, 10 ML
SDP
Catalog No. NC2887465
Encompass_Preferred
Fast DNA extraction kit provides a simple and rapid method to isolate the genomic DNA from tissues or cells for PCR amplification, genotype identification, CRISPR mutation detection, and library screening.
Catalog No. NC2887465 Supplier TRIBIOSCIENCE INC Supplier No. TBS600810
Fast DNA extraction kit provides a simple and rapid method to isolate the genomic DNA from tissues or cells for PCR amplification, genotype identification, CRISPR mutation detection, and library screening.