Learn More
Alkali Scientific Genomic DNA Extraction Spin Co
SDP
Catalog No. 503027649
Encompass_Preferred
Alkali ScientificDNA Spin Columns are designed for the rapid reliable and high-quality extraction of DNA from a wide range of biological samples including tissues cells plants and blood Constructed from durable polypropylene these spin columns ensure chemical compatibility and sample integrity while their advanced silica membrane technology facilitates efficient DNA binding washing and elution Supplied with collection tubes they offer convenience and ease of use for researchers across various applications These columns deliver consistent and reproducible yields of DNA with large fragment sizes and high purity making them suitable for applications such as PCR qPCR cloning sequencing and genotyping Their DNase- and RNase-free construction along with human DNA-free certification ensures contamination-free results for sensitive workflows Polypropylene Construction Ensures durability chemical resistance and sample integrity High Purity DNA Delivers large fragment sizes suitable for ad
Catalog No. 50-302-7649 Supplier Alkali Scientific Supplier No. GE9002
Add to Cart
Add to Cart
Alkali ScientificDNA Spin Columns are designed for the rapid reliable and high-quality extraction of DNA from a wide range of biological samples including tissues cells plants and blood Constructed from durable polypropylene these spin columns ensure chemical compatibility and sample integrity while their advanced silica membrane technology facilitates efficient DNA binding washing and elution Supplied with collection tubes they offer convenience and ease of use for researchers across various applications These columns deliver consistent and reproducible yields of DNA with large fragment sizes and high purity making them suitable for applications such as PCR qPCR cloning sequencing and genotyping Their DNase- and RNase-free construction along with human DNA-free certification ensures contamination-free results for sensitive workflows Polypropylene Construction Ensures durability chemical resistance and sample integrity High Purity DNA Delivers large fragment sizes suitable for ad